Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Human Recombinant UBC3B (from E. coli)

Human Recombinant UBC3B (from E. coli)

Catalog # ABCAAB173060-10
Marketsource
Supplier:   Abcam
Human Recombinant UBC3B (from E. coli)
Catalog # ABCAAB173060-10
Supplier:  Abcam
Marketsource
CAS Number:  
Human Recombinant UBC3B (from E. coli)
Human Recombinant UBC3B (from E. coli)
CAS Number:  
Price
140.00 SFr.
Log in for availability
1 * 10 µG
Product Details & Documents

Recombinant Human UBC3B protein is a Human Full Length protein, in the 1 to 238 aa range, expressed in Escherichia coli, with >95% purity, <1 EU/µg endotoxin level and suitable for SDS-PAGE, HPLC.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Specifications

Specifications

  • Konjugation:
    Unconjugated
  • Form:
    Liquid
  • Transporttemperatur:
    Dry Ice
  • Validierte Anwendungen:
    HPLC, SDS-PAGE
  • Purity:
    >95%
  • Spezies:
    Human
  • Biologische Aktivität:
    Standard
  • Zusammensetzung:
    pH: 7,5, Constituents: 1,19% HEPES, 0,88% Sodium chloride
  • Cat. No.:
    ABCAAB173060-10
  • Ursprung:
    E. coli
  • Endotoxingehalt:
    <1 EU/µg
  • Tierfrei:
    N
  • Protein/Peptid-Typ:
    Rekombinant
  • Lagerbedingungen:
    Store at −20 °C
  • Protein/Peptid-Name:
    UBC3B
  • Proteinsynonym:
    CDC34B|UBE2R2|Ubiquitin-conjugating enzyme E2 R2|E2 ubiquitin-conjugating enzyme R2|Ubiquitin carrier protein R2|Ubiquitin-conjugating enzyme E2-CDC34B|Ubiquitin-protein ligase R2

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.