Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Mouse Recombinant TIM 3 (from HEK293 cells)

Mouse Recombinant TIM 3 (from HEK293 cells)

Catalog # ABCAAB316525-10UG
New Product
Supplier:   Abcam
Mouse Recombinant TIM 3 (from HEK293 cells)
Catalog # ABCAAB316525-10UG
Supplier:  Abcam
New Product
CAS Number:  
Mouse Recombinant TIM 3 (from HEK293 cells)
Mouse Recombinant TIM 3 (from HEK293 cells)
CAS Number:  
Price
SFr. 185.00
Log in for availability
1 * 10 µG
Product Details & Documents

Recombinant Mouse TIM-3 Protein Standard (His tag) is a Mouse Fragment protein, expressed in HEK 293, with ≥80% purity and suitable for sELISA, SDS-PAGE.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Specifications

Specifications

  • Konjugation:
    Unconjugated
  • Form:
    Liquid
  • Transporttemperatur:
    Dry Ice
  • Validierte Anwendungen:
    SDS-PAGE, sELISA
  • Purity:
    ≥80%
  • Spezies:
    Mouse
  • Biologische Aktivität:
    Standard
  • Zusammensetzung:
    pH: 7,3 - 7,5, Constituents: 2,922% Sodium chloride, 0,64107% disodium;hydrogen phosphate;dodecahydrate, 0,02858% Potassium phosphate monobasic
  • Cat. No.:
    ABCAAB316525-10UG
  • Ursprung:
    HEK293 cells
  • Tierfrei:
    Y
  • Protein/Peptid-Typ:
    Rekombinant
  • Lagerbedingungen:
    Store at −80 °C
  • Protein/Peptid-Name:
    TIM 3
  • Proteinsynonym:
    CD366|Tim3|Timd3|Havcr2|Hepatitis A virus cellular receptor 2 homolog|HAVcr-2|T-cell immunoglobulin and mucin domain-containing protein 3|T-cell immunoglobulin mucin receptor 3|T-cell membrane protein 3|TIMD-3|TIM-3

Specifications