Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Human Native Factor IX/PTC | Avantor

Human Native Factor IX/PTC

Katalognr. ABCAAB62544-100
Marketsource
Anbieter:   Abcam
Human Native Factor IX/PTC
Katalognr. ABCAAB62544-100
Anbieter:  Abcam
Marketsource
CAS Nummer:  
Human Native Factor IX/PTC
Human Native Factor IX/PTC
CAS Nummer:  
Preis
SFr. 275.00
Einloggen für Verfügbarkeit
1 * 100 µG
Product Details & Documents

Native Human Factor IX/PTC protein is a Human Full Length protein, expressed in Native, with >95% purity and suitable for SDS-PAGE, FuncS.

Achtung! Bitte beachten: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Technische Daten

Technische Daten

  • Cat. No.:
    ABCAAB62544-100
  • Tierfrei:
    N
  • Konjugation:
    Unconjugated
  • Validierte Anwendungen:
    FuncS, SDS-PAGE
  • Purity:
    >95%
  • Spezies:
    Human
  • Protein/Peptid-Typ:
    Nativ
  • Lagerbedingungen:
    −20 °C
  • Protein/Peptid-Name:
    Factor IX/PTC
  • Proteinsynonym:
    Coagulation factor IX|Christmas factor|Plasma thromboplastin component|PTC|F9

Technische Daten

Recommandations

Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".

Sinon, vous recevrez des recommandations génériques.