Human ACE2 ELISA Kit
Human ACE2 ELISA kit, 96 tests
Katalognr. ABCAAB235649-96
Anbieter: Abcam
Direkt zu
Product Details & Documents
Human ACE2 ELISA Kit is a single-wash 90-min Simplestep used to quantify Human ACE2 with a sensitivity of 1052 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader. - Cited in over 20 citations.
Technische Daten
Technische Daten
- Bezeichnung:Human ACE2 ELISA kit
- Tests pro Kit:96 tests
- Empfindlichkeit:1052 pg/ml
- Reaktivität der primären Antikörper:Human
- Testdauer:One step
- Time to Results:1 h 30 min
- Cat. No.:ABCAAB235649-96
- Lagertemperatur:+4 °C
- Format:Pre-coated
- Target protein:ACE2
- Erkennungsmethode:Colorimetric
- Testtyp:Sandwich
- Probentyp:Urine, Tissue Extracts, Cell culture media, Heparin Plasma, Citrate plasma, Serum, EDTA Plasma
- Erfassungsbereich:3,98 - 255 ng/ml
- Regulatory status:RUO



