Human IL-8 ELISA Kit
Human IL-8 ELISA kit is a single-wash, 90-minute SimpleStep ELISA® used to quantify human interleukin-8 (IL-8) with a sensitivity of 1,8 pg/ml. The assay employs a simple Mix-Wash-Read protocol with just one incubation and wash step. - Colorimetric sandwich ELISA - 450 nm readout; compatible with any standard plate reader - Available in different formats to meet various throughput needs, including 384-well plates for higher throughput - Broad sample compatibility – suitable for serum, plasma, cell culture supernatants, and cerebrospinal fluid - Cited in over 30 publications.
Human IL-8 ELISA kit is a single-wash, 90-minute SimpleStep ELISA® used to quantify human interleukin-8 (IL-8) with a sensitivity of 1,8 pg/ml. The assay employs a simple Mix-Wash-Read protocol with just one incubation and wash step. - Colorimetric sandwich ELISA - 450 nm readout; compatible with any standard plate reader - Available in different formats to meet various throughput needs, including 384-well plates for higher throughput - Broad sample compatibility – suitable for serum, plasma, cell culture supernatants, and cerebrospinal fluid - Cited in over 30 publications.
- Catalog No:
- ABCAAB214030-96
- ABCAAB214030-10X96
- ABCAAB214030-1X384
- Target protein:
- IL-8
- IL-8
- IL-8
- Reaktivität der primären Antikörper:
- Human
- Human
- Human
- Testtyp:
- Sandwich
- Sandwich
- Sandwich
- Format:
- Pre-coated
- Pre-coated
- Pre-coated
- Testdauer:
- One step
- One step
- One step
- Empfindlichkeit:
- 1,8 pg/ml
- 1,8 pg/ml
- 1,8 pg/ml
- Erfassungsbereich:
- 3,91 - 250 pg/ml
- 3,91 - 250 pg/ml
- 3,91 - 250 pg/ml
- Erkennungsmethode:
- Colorimetric
- Colorimetric
- Colorimetric
- Probentyp:
- Cerebral Spinal Fluid, Heparin Plasma, Citrate plasma, Cell culture supernatant, Serum, EDTA Plasma
- Cerebral Spinal Fluid, Heparin Plasma, Citrate plasma, Cell culture supernatant, Serum, EDTA Plasma
- Cerebral Spinal Fluid, Heparin Plasma, Citrate plasma, Cell culture supernatant, Serum, EDTA Plasma
- Time to Results:
- 1 h 30 min
- 1 h 30 min
- 1 h 30 min
- Regulatory status:
- RUO
- RUO
- RUO
- Lagertemperatur:
- 4 °C
- 4 °C
- 4 °C
Product Family Options
- BezeichnungTests pro Kit
- Human IL-8 ELISA kit96 tests1 * 96 TestsSFr. 769.00
Specifications
Cat. No.ABCAAB214030-96Target proteinIL-8Reaktivität der primären AntikörperHumanTesttypSandwichFormatPre-coatedTestdauerOne stepEmpfindlichkeit1,8 pg/mlErfassungsbereich3,91 - 250 pg/mlErkennungsmethodeColorimetricProbentypCerebral Spinal Fluid, Heparin Plasma, Citrate plasma, Cell culture supernatant, Serum, EDTA PlasmaTime to Results1 h 30 minRegulatory statusRUOLagertemperatur4 °C - Human IL-8 ELISA kit10×96 tests1 * 960 TestsSFr. 6’930.00
Specifications
Cat. No.ABCAAB214030-10X96Target proteinIL-8Reaktivität der primären AntikörperHumanTesttypSandwichFormatPre-coatedTestdauerOne stepEmpfindlichkeit1,8 pg/mlErfassungsbereich3,91 - 250 pg/mlErkennungsmethodeColorimetricProbentypCerebral Spinal Fluid, Heparin Plasma, Citrate plasma, Cell culture supernatant, Serum, EDTA PlasmaTime to Results1 h 30 minRegulatory statusRUOLagertemperatur4 °C - Human IL-8 ELISA kit384 tests1 * 1 TestsSFr. 1’900.00New Product
Specifications
Cat. No.ABCAAB214030-1X384Target proteinIL-8Reaktivität der primären AntikörperHumanTesttypSandwichFormatPre-coatedTestdauerOne stepEmpfindlichkeit1,8 pg/mlErfassungsbereich3,91 - 250 pg/mlErkennungsmethodeColorimetricProbentypCerebral Spinal Fluid, Heparin Plasma, Citrate plasma, Cell culture supernatant, Serum, EDTA PlasmaTime to Results1 h 30 minRegulatory statusRUOLagertemperatur4 °CNew Product



