Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
mCherry ELISA Kit

mCherry ELISA Kit

mCherry ELISA kit, 96 tests
Catalog # ABCAAB221829-96
Supplier:   Abcam
mCherry ELISA Kit
Catalog # ABCAAB221829-96
Supplier:  Abcam
CAS Number:  
mCherry ELISA Kit
mCherry ELISA Kit

mCherry ELISA kit, 96 tests

CAS Number:  
Price
SFr. 828.00
Log in for availability
1 * 96 Tests
Product Details & Documents

mCherry ELISA Kit is a single-wash 90-min SimpleStep Sandwich ELISA used to quantify mCherry with a sensitivity of 0,47 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric assay - 450 nm readout, works on any plate reader - Validated in cell culture extracts, cell culture media and cell culture supernatant.

Specifications

Specifications

  • Bezeichnung:
    mCherry ELISA kit
  • Tests pro Kit:
    96 tests
  • Empfindlichkeit:
    0,47 pg/ml
  • Reaktivität der primären Antikörper:
    Species independent
  • Testdauer:
    One step
  • Time to Results:
    1 h 30 min
  • Cat. No.:
    ABCAAB221829-96
  • Lagertemperatur:
    +4 °C
  • Format:
    Pre-coated
  • Target protein:
    mCherry
  • Erkennungsmethode:
    Colorimetric
  • Testtyp:
    Sandwich
  • Probentyp:
    Cell culture extracts, Cell culture media, Cell culture supernatant
  • Erfassungsbereich:
    6,25 - 400 pg/ml
  • Regulatory status:
    RUO

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.