Human Trypsin ELISA Kit
Human Trypsin ELISA kit, 96 tests
Katalognr. ABCAAB277474-1X96
Anbieter: Abcam
Direkt zu
Product Details & Documents
Human Trypsin ELISA kit is a Sandwich (quantitative) ELISA kit for the measurement of Human Trypsin in Human in plasma, cell culture supernatant, Serum samples.
Technische Daten
Technische Daten
- Bezeichnung:Human Trypsin ELISA kit
- Tests pro Kit:96 tests
- Cat. No.:ABCAAB277474-1X96
- Lagertemperatur:4 °C
- Format:Pre-coated
- Target protein:Trypsin
- Erkennungsmethode:Colorimetric
- Empfindlichkeit:0,29 ng/ml
- Testtyp:Sandwich
- Probentyp:Plasma, cell culture supernatant, serum
- Reaktivität der primären Antikörper:Human
- Erfassungsbereich:0,274 - 200 ng/ml
- Testdauer:Multiple steps
- Regulatory status:RUO



