Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Cat AGP ELISA Kit

Cat AGP ELISA Kit

Cat AGP ELISA kit, 96 tests
Catalog # ABCAAB291068-1X96
Supplier:   Abcam
Cat AGP ELISA Kit
Catalog # ABCAAB291068-1X96
Supplier:  Abcam
CAS Number:  
Cat AGP ELISA Kit
Cat AGP ELISA Kit

Cat AGP ELISA kit, 96 tests

CAS Number:  
Price
759.00 SFr.
Log in for availability
1 * 96 Tests
Product Details & Documents

Cat AGP ELISA kit is a ELISA kit for the measurement of Cat AGP in Cat in plasma, Serum samples.

Specifications

Specifications

  • Bezeichnung:
    Cat AGP ELISA kit
  • Tests pro Kit:
    96 tests
  • Cat. No.:
    ABCAAB291068-1X96
  • Lagertemperatur:
    4 °C
  • Format:
    Pre-coated
  • Target protein:
    Alpha 1 Acid Glycoprotein/AGP
  • Testtyp:
    Sandwich
  • Probentyp:
    Plasma, serum
  • Reaktivität der primären Antikörper:
    Cat
  • Erfassungsbereich:
    1,56 - 100 ng/ml
  • Testdauer:
    Multiple steps
  • Regulatory status:
    RUO

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.