SimpleStep ELISA® Custom ELISA Kit
ABCAAB270552-5X96
: Abcam
:
- Bezeichnung:SimpleStep ELISA® Custom ELISA kit
- Tests pro Kit:5×96 tests
SimpleStep ELISA® Custom ELISA kit allows you to develop your own SimpleSte ELISA® using any antibody pair. - No plate coating needed - Possibility to measure different target on the same plate - All components included, just add the antibody pair - Streamline your ELISA® to a single-wash format if desired