Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Mouse MBL ELISA Kit | Avantor

Mouse MBL ELISA Kit

Mouse MBL ELISA kit, 96 tests
Katalognr. ABCAAB208343-96
Anbieter:   Abcam
Mouse MBL ELISA Kit
Katalognr. ABCAAB208343-96
Anbieter:  Abcam
CAS Nummer:  
Mouse MBL ELISA Kit
Mouse MBL ELISA Kit

Mouse MBL ELISA kit, 96 tests

CAS Nummer:  
Preis
SFr. 749.00
Einloggen für Verfügbarkeit
1 * 96 Tests
Product Details & Documents

Mouse MBL ELISA kit is a single-wash 90-min Simplestep used to quantify Mouse MBL with a sensitivity of 8,5 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric sandwich ELISA - 450 nm readout : works on any standard plate reader - Design your own immunoassay: we also offer the conjugation-ready antibody pair.

Zertifizierung: RUO

Technische Daten

Technische Daten

  • Bezeichnung:
    Mouse MBL ELISA kit
  • Tests pro Kit:
    96 tests
  • Empfindlichkeit:
    8,5 pg/ml
  • Reaktivität der primären Antikörper:
    Mouse
  • Testdauer:
    One step
  • Time to Results:
    1 h 30 min
  • Cat. No.:
    ABCAAB208343-96
  • Lagertemperatur:
    4 °C
  • Format:
    Pre-coated
  • Target protein:
    Mannan Binding Lectin/MBL
  • Erkennungsmethode:
    Colorimetric
  • Testtyp:
    Sandwich
  • Probentyp:
    Plasma, Cell culture extracts, Tissue Extracts, Cell culture supernatant, Serum
  • Erfassungsbereich:
    39,063 - 2500 pg/ml
  • Regulatory status:
    0

Technische Daten

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.