AuftragsverfolgungKontakt
Human Tissue Factor ELISA Kit
Human Tissue Factor ELISA Kit
Human Tissue Factor ELISA kit, 96 tests
Catalog # ABCAAB220653-96
Supplier:   Abcam
Human Tissue Factor ELISA Kit
Catalog # ABCAAB220653-96
Supplier:  Abcam
CAS Number:  
Human Tissue Factor ELISA Kit
Human Tissue Factor ELISA Kit
Human Tissue Factor ELISA kit, 96 tests
CAS Number:  
Price
774.00 SFr.
Log in for availability
1 * 96 Tests
Product Details & Documents
Human Tissue Factor ELISA Kit is a single-wash 90-min Simplestep used to quantify Human Tissue Factor with a sensitivity of 3,6 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader. - Cited in over 10 citations.
Specifications
Specifications
  • Bezeichnung:
    Human Tissue Factor ELISA kit
  • Tests pro Kit:
    96 tests
  • Empfindlichkeit:
    3,6 pg/ml
  • Reaktivität der primären Antikörper:
    Human
  • Testdauer:
    One step
  • Time to Results:
    1 h 30 min
  • Cat. No.:
    ABCAAB220653-96
  • Lagertemperatur:
    +4 °C
  • Format:
    Pre-coated
  • Target protein:
    Tissue Factor
  • Erkennungsmethode:
    Colorimetric
  • Testtyp:
    Sandwich
  • Probentyp:
    Saliva, Urine, Cell culture extracts, Heparin Plasma, Citrate plasma, Cell culture supernatant, EDTA Plasma
  • Erfassungsbereich:
    15,63 - 1000 pg/ml
  • Regulatory status:
    RUO
Specifications