Pipette tips for Eppendorf®
Catalog # GREI765280UK
Anbieter: Greiner Bio-One
Technische Daten
- Verpackt:96 per ST Rack (yellow)
- Steril:-
- Volumen:0,5 - 20 µl
- Länge:45,6 mm
- Seq:04
- Spitzentyp:Micro, crystal tip, UK
- Spitzenfarbe:Transparent
- Zur Nutzung mit:Eppendorf® 4710, Reference, Research Pro
- VE:960
Technische Daten
Über diesen Artikel
These pipette tips feature a shorter tip collar and are recommended for use with Eppendorf® pipettors.
- Bevelled edge increases pipetting accuracy
- Manufactured from high-grade PP to give the optimum in performance and fit
- Free of heavy metals
- Autoclavable
Ordering information: Matching racks for bulkware tips can be ordered separately.