PCR work-up racks, 96-well
Anbieter: Greiner Bio-One
Zertifikate
Über diesen Artikel
Suitable for 0,2 ml reaction tubes and tube strips.
- Alphanumerical coding for position
- Can be firmly located in the PCR workstation
- Thermal resistance down to −80 °C
Frequently Bought Together