ContactUsLinkComponent
Special marker pens
Catalog # GREI841077UK
Supplier:  Greiner Bio-One
Special marker pens
Catalog # GREI841077UK
Supplier:  Greiner Bio-One
Specifications
  • Steril:
    -
  • Farbe:
    Schwarz
  • Bezeichnung:
    Spezieller Markerstift, sehr feine Dicke
  • Seq:
    02
  • VE:
    1
Specifications
About this item
Marker pens designed for labelling all plastic articles and glassware.
  • Water-resistant
  • Quick drying
Ordering information: Available in four colours and two writing thicknesses.