Test tubes,with rim
70 mm, 7,8 mm, 0,45 mm
Catalog # HECH42771015
Supplier: GLASWARENFABRIK KARL HECHT
Jump to:
Product Details & Documents
FIOLAX®, neutral glass.
- Very good chemical resistance
- Relatively resistant against rapid temperature changes and local heating
Thin walled test tubes for a wide variety of applications.
Specifications
- Ext.Ø:7,8 mm
- Wandstärke:0,45 mm
- Länge:70 mm