Instruments/Medicine trays, with Plexiglas® stand
Catalog # HECH41105010
Supplier: GLASWARENFABRIK KARL HECHT
Specifications
- Typ:with 20 holes
- VE:1
Specifications
About this item
Made of melamin/Plexiglas®
- Each row of holes has an area for inscription (pencil notes can easily be removed by rubber or moist cloth)
- The free space at the side is provided for other medicines, instruments etc.
- Without cups
Consisting of plastic tray of melamine, white, rack of plexiglas® for 20 medicine cups.