Transfection reagent, TurboFect in-vitro
Katalognr. FERMR0531
Anbieter: Thermo Fisher Scientific
CAS-Nummer:
Technische Daten
- VE:1 KIT
- DNA:Ja
- Bezeichnung:TurboFect in-vitro Transfektionsreagenz, 1 ml
Technische Daten
Über diesen Artikel
TurboFect transfection reagent is recommended for efficient transfection of plasmid DNA. Distinct from lipid-based formulas, TurboFect is a sterile solution of a proprietary cationic polymer in water. The polymer forms positively-charged complexes with DNA that are both compact and stable. These complexes protect DNA from degradation and facilitate efficient plasmid delivery into eukaryotic cells.
- High transfection efficiency of plasmid DNA into a wide variety of cell types (adherent and suspension cells; differentiated and undifferentiated cells)
- Excellent transfection efficiency in the presence or absence of serum
- Ready to use reagent – no need to reconstitute, dilute, or manipulate
- Simple protocol