Pycnometer for ice creme tests
Katalognr. WITG3.935.003
Anbieter: witeg Labortechnik
Technische Daten
- Volumen (nom.):20 ml
- Environmentally Preferable:
- VE:1
Technische Daten
Über diesen Artikel
Pycnometer, made of borosilicate glass 3.3.
- Cap. 20 %
- Nominal capacity printed on the bottom
- Very good chemical and temperature resistance