Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Proteine und Peptide
Soy bean trypsin inhibitor
Soy bean trypsin inhibitor
Soy bean trypsin inhibitor
Catalog # CHMCAB1239
Supplier:
Chemicon
CAS Number:
Expand image
Soy bean trypsin inhibitor
Catalog # CHMCAB1239
Supplier:
Chemicon
CAS Number:
Specifications
VE:
25 mg
CAS Nr.:
9035-81-8
Spezies:
Soy bean
Lagerbedingungen:
Freezer
Protein/Peptid-Name:
Trypsin inhibitor
Specifications
About this item
Product Details
Product Details