Order Entry
Switzerland
ContactUsLinkComponent
 
Leupeptin hemisulphate
  CHMCEI8
 :  Chemicon
 :  
Leupeptin hemisulphate
  CHMCEI8
 :  Chemicon
 :  

 

  • VE:
    100 mg
  • CAS Nr.:
    103476-89-7
  • Lagerbedingungen:
    Freezer
  • MDL No.:
    MFCD00037012
  • Proteinsynonym:
    (Ac-Leu-Leu-Arg-al hemisulphate) monohydrate
  • Protein/Peptid-Name:
    Leupeptin hemisulphate
  • Molecular formula:
    C₄₀H₇₈N₁₂O₁₂S