
Amidoschwarz 10 B (Naphtholblauschwarz, Säureschwarz 1)
Catalog # NADIHS-601-25G
Supplier: NATIONAL DIAGNOSTICS
CAS Number:
Specifications
- Packung:Glass bottle for solids
- VE:25 g
- Seq:3308
Specifications
About this item
:
:616.49 g/mol
EINECS:213-903-1
:GHS07
Storage Temperature:Raumtemperatur
CAS Number:1064-48-8
Formula:C₂₂H₁₄N₆Na₂O₉S₂
MDL Number:MFCD00004017