Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Chymostatin

Chymostatin

100 mg
  CHMCEI6
 :   Chemicon
Chymostatin
  CHMCEI6
 :  Chemicon
 :  
Chymostatin

100 mg

 :  
 
SFr. 3’890.00
   
1 * 100 mg
 
 

 

  • VE:
    100 mg
  • Cat. No.:
    CHMCEI6
  • CAS Nr.:
    9076-44-2
  • MDL No.:
    MFCD00071059
  • Lagerbedingungen:
    Freezer
  • Protein/Peptid-Name:
    Chymostatin

 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.