SP Bel-Art® tapered plug sampler
Catalog # BELAH378260000
Supplier: Bel-Art Products, a Part of SP
Specifications
- Ø:17 mm
- Bezeichnung:Tapered plug sampler
- Länge:191 mm
- Material:Edelstahl
- VE:1
Specifications
About this item
Tapered plug sampler can remove a sample from bags.
- Sharp forward edge pierces packaging to retrieve sample
- T-shape handle applies rotating force to cut sample
- Easy cleaning and corrosion resistance
Suitable for blocks of cheese, grease, soap, clay, or any other similar semi-solids.