Ampicillin Trihydrat
Glass bottle, 500 ml
Katalognr. CHMCS4601
Anbieter: Chemicon
Direkt zu
Product Details & Documents
Synonyme: (2S,5R,6R)-6-((R)-2-Amino-2-phenylacetamido)-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid trihydrate
- CAS Nummer:7177-48-2
- Formel:C₁₆H₁₉N₃O₄S·3H₂O
- MDL-Nummer:MFCD00072036
- Molekulargewicht:403.45 g/mol
- Lagertemperatur:Kühlschrank
- EINECS:200-709-7
Technische Daten
Technische Daten
- Packung:Glass bottle
- VE:500 ml
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



