Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Bis(2-chlorethyl)ammoniumchlorid

Bis(2-chlorethyl)ammoniumchlorid

Glass bottle for solids, 500 g
Catalog # MOLE90037967
Marketsource
Supplier:   Molekula
Bis(2-chlorethyl)ammoniumchlorid
Catalog # MOLE90037967
Supplier:  Molekula
Marketsource
CAS Number:  821-48-7
Bis(2-chlorethyl)ammoniumchlorid

Glass bottle for solids, 500 g

CAS Number:  821-48-7
Price
Inquire For Price
Log in for availability
1 * 500 g
Product Details & Documents
Synonyms:  Bis(2-chloroethyl)amine hydrochloride
  • CAS Number:821-48-7
  • Formula:C₄H₁₀Cl₃N
  • MDL Number:MFCD00012515
  • Molecular Weight:178.49 g/mol
  • EINECS:212-479-5
  • UN:2923
  • ADR:8,II
Specifications

Specifications

  • Packung:
    Glass bottle for solids
  • VE:
    500 g

Specifications