Bosshead, SP Bel-Art
Catalog # BELAH183500000
Supplier: Bel-Art Products, a Part of SP
Specifications
- Material:Edelstahl
- VE:1
- Für Stab Ø:13 mm
Specifications
About this item
Stainless steel. This open-sided bosshead accepts rods up to 13 mm diameter.
Holds rods at right angles to one another with flat wing-head clamping screws.