Probenehmer für Säuren, Precisionware
Probenehmer für Chemikalien, Weiß, PP, 500 ml, 142 cm
Katalognr. BELAH378010000
Anbieter: Bel-Art Products, a Part of SP
Probenehmer für Säuren, Precisionware
Katalognr. BELAH378010000
Anbieter: Bel-Art Products, a Part of SP
Direkt zu
Product Details & Documents
PE bottle threads on to end of rod to collect samples. Safely distances user from dangerous chemicals.
- Fluid enters through fitting above bottle
- T-handle for secure grip when lowering sampler
- Handle has two piece threaded joint to shorten during transport or storage
Designed for hydrochloric, sulfuric and acetic acids, formaldehyde potassium or sodium hydroxide, in tanks rail cars, and other large vessels.
Technische Daten
- Kapazität:500 ml
- Farbe:Weiß
- Bezeichnung:Probenehmer für Chemikalien
- Länge:142 cm
- Material:PP