Deckglaspinzetten
Gebogen, 102 mm
Katalognr. BELAH379420000
Anbieter: Bel-Art Products, a Part of SP
Direkt zu
Product Details & Documents
These PTFE coated forceps with wide tips are perfect for lifting cover glasses.
- Available in two styles: With bent or straight tip
- FDA compliant for food contact
Technische Daten
- Typ:Gebogen
- Länge:102 mm
- Seq:01