Order Entry
Switzerland
ContactUsLinkComponent
 
Chlorflurenol-Methylester (2-Chlor-9-hydroxy-9H-fluoren-9-carbonsäuremethylester)
Catalog # EHERC11305000
Supplier:  EHRENSTORFER
CAS Number:  
Chlorflurenol-Methylester (2-Chlor-9-hydroxy-9H-fluoren-9-carbonsäuremethylester)
Catalog # EHERC11305000
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Packung:
    Glass bottle for solids
  • VE:
    0,25 g
  • Seq:
    3029

Specifications

About this item

 :
 :274.7 g/mol
EINECS:219-800-8
CAS Number:2536-31-4
Formula:C₁₅H₁₁ClO₃