TLC spray cabinets
CAMA022.6230UK
: CAMAG
- Bezeichnung:TLC spray cabinet II, UK-plug, 470×490×490 mm, with blower and 1,5 m flexible exhaust hose (127 mm Ø)
- Seq:02
- VE:1
The TLC spray cabinet is designed for the complete removal of excessive spray mist while spraying a TLC plate with reagent.
- Made of PVC
- The blower, a radial fan driven by a motor outside of the fume duct, produces an airflow of 3,7 m³/min
- Removable built in tray, easy for cleaning
There is no deflection of the spray jet before it reaches the plate, an effect often encountered in a normal laboratory fume hood. Particles rebounding from the plate are completely removed. This spray cabinet is also useful for drying plates after development, with or without the assistance of a hair dryer.