Magnetic stirring bar set, with retriever, BOLA
Catalog # BOHLC348-10
Supplier: Bohlender
Specifications
- Typ:Magnetic stirring bar set
- VE:1 SET
Specifications
About this item
Magnetic stirring bar set containing the most common magnetic stirring bars and a retriever.
- Chemical resistant
- Ideal for beginners, for testing different kinds and dimensions of magnetic stirring bars
Temperature resistance: −200 to +250 °C.
L×W×H of box: 175×110×30 mm
Delivery: Set consists of one each of (lengthר): Cylindrical 10×6, 15×4,5, 20×6, 25×6, 30×6, 40×8, 50×8, 60×9 mm; Pivot ring 15×5, 25×6, 40×8 mm; Triangular 25×8, 40×14 mm; and 150 mm retriever.