Magnetic stirring bars, PTFE coated, ball shaped
KART770
: KARTELL
- Ø:12,57 mm
- VE:1
Magnetic stirrers which are ideal for test tubes or for eccentric stirring movements.
- Suitable for use with foodstuffs
Magnetic stirrers which are ideal for test tubes or for eccentric stirring movements.