Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
 

Silbernitrat

Oxidizing
Corrosive
Environment
Supplier:   EUROCLONE
Silbernitrat
Supplier:  EUROCLONE
Certificates
Silbernitrat

4 Options Available Below

Product Details & Documents
Synonyms:  Lunar caustic, Silver mononitrate, Nitric acid silver salt
  • CAS Number:7761-88-8
  • Formula:AgNO₃
  • MDL Number:MFCD00003414
  • Molecular Weight:169.87 g/mol
  • Merck Index:12,08661
  • Boiling Pt:444 °C (1013 hPa)
  • Melting Pt:210 °C
  • Density:4,35 g/cm³ (20 °C)
  • Storage Temperature:Raumtemperatur
  • EINECS:231-853-9
  • UN:1493
  • ADR:5.1,II
  • REACH:01-2119513705-43
Product Family Options

Product Information

  • VE
    Availability
    Price
  • 1 KIT
    Log in for availability
    Log in for availability
    1 * 1 KIT
    1 * 1 KIT

  • 1 KIT
    Log in for availability
    Log in for availability
    1 * 1 KIT
    1 * 1 KIT

  • 50 g
    Log in for availability
    Log in for availability
    1 * 50 g
    1 * 50 g

  • 250 g
    Log in for availability
    Log in for availability
    1 * 250 g
    1 * 250 g

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.