Order Entry
Switzerland
ContactUsLinkComponent
Natriumbenzolsulfinat
Natriumbenzolsulfinat
Catalog # MOLEM34253542
Supplier:  Molekula
CAS Number:  
Natriumbenzolsulfinat
Catalog # MOLEM34253542
Supplier:  Molekula
CAS Number:  

Specifications

  • Packung:
    Glass bottle for solids
  • VE:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :164.16 g/mol
EINECS:212-842-8
CAS Number:873-55-2
Formula:C₆H₅NaO₂S
MDL Number:MFCD00013135