2-Methyl-2-thiazolin
Glasflasche, 10 ml
Anbieter: Merck
Direkt zu
Product Details & Documents
Synonyme: 2-Methyl-4,5-dihydrothiazole
- CAS Nummer:2346-00-1
- Formel:C₄H₇NS
- MDL-Nummer:MFCD00005314
- Molekulargewicht:101.17 g/mol
- EINECS:219-071-6
Documents
msds_814232
SDS
Technische Daten
Ergebnisse des Spezifikationstests
- Solubility:(20 °C) soluble
- Melting Point:-101 °C
- Molar Mass:101.17 g/mol
- Boiling Point:144 - 146 °C (1013 hPa)
- Flash Point:37 °C
- Refractive Index:1.5211 (20 °C, 589 nm)
- Density:1.08 g/cm3 (20 °C)
Technische Daten
- Packung:Glasflasche
- VE:10 ml
- Cat. No.:8.14232.0010
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



