Centrifuge tubes
Anbieter: MARIENFELD
Direkt zu
Über diesen Artikel
Soda lime glass or borosilicate glass 3,3.
- Graduated or ungraduated
- With beaded or straight rim
- Good chemical resiostance
Long conical type.
4 Varianten
Product Details & Documents
Soda lime glass or borosilicate glass 3,3.
- Graduated or ungraduated
- With beaded or straight rim
- Good chemical resiostance
Long conical type.
Product Family Options
Soda lime glass
Produktinformationen
- KapazitätØ×LVerschlussartTypVerpacktVerfügbarkeitPreis
- 15 ml~16/17×112/113 mmungraduated, beaded rim100/BoxEinloggen für Verfügbarkeit1 * 100 ST113.00 SFr.
- 15 ml~16/17×112/113 mmOhne Verschlusswhite graduated 1 to 10 : 0.1 ml, with mark at 15 ml, beaded rim100/BoxEinloggen für Verfügbarkeit1 * 100 ST306.00 SFr.
- 15 ml~16/17×112/113 mmOhne Verschlusswhite graduated 1 to 15 : 0.1 ml, beaded rim100/BoxEinloggen für Verfügbarkeit1 * 100 ST306.00 SFr.
- 15 ml~16/17×112/113 mmOhne Verschlussgraduated, straight rim100/BoxEinloggen für Verfügbarkeit1 * 100 ST306.00 SFr.