Natriumhydroxid
Anbieter: EUROCLONE
Direkt zu
Über diesen Artikel
Synonyme: Caustic soda
- CAS Nummer:1310-73-2
- Formel:NaOH
- MDL-Nummer:MFCD00003548
- Molekulargewicht:40 g/mol
- Merck-Index:14,08627
- Siedepunkt:1390 °C (1013 hPa)
- Schmelzender Platingehalt:323 °C
- Dichte:2,13 g/cm³ (20 °C)
- Lagertemperatur:Raumtemperatur
- EINECS:215-185-5
- UN:1823
- ADR:8,II
- ERREICHEN:01-2119457892-27
2 Varianten
Product Details & Documents
Synonyme: Caustic soda
- CAS Nummer:1310-73-2
- Formel:NaOH
- MDL-Nummer:MFCD00003548
- Molekulargewicht:40 g/mol
- Merck-Index:14,08627
- Siedepunkt:1390 °C (1013 hPa)
- Schmelzender Platingehalt:323 °C
- Dichte:2,13 g/cm³ (20 °C)
- Lagertemperatur:Raumtemperatur
- EINECS:215-185-5
- UN:1823
- ADR:8,II
- ERREICHEN:01-2119457892-27
Product Family Options
Produktinformationen
- VEVerfügbarkeitPreis
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



