Order Entry
Switzerland
ContactUsLinkComponent
Natriumcyantrihydroborat
Flammable
Toxic
Corrosive
Environment
Natriumcyantrihydroborat
Catalog # MOLE23955696-25G
Supplier:  Molekula
CAS Number:  
Flammable
Toxic
Corrosive
Environment
Natriumcyantrihydroborat
Catalog # MOLE23955696-25G
Supplier:  Molekula
CAS Number:  

Specifications

  • Packung:
    Glass bottle for solids
  • VE:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :62.84 g/mol
Merck Index:13,08680
Melting Pt:241…243 °C
Density:1.11…1.12 g/cm³ (28 °C)
Storage Temperature:Raumtemperatur
EINECS:247-317-2
UN:3134
ADR:4.3,II
 :GHS02
 :GHS06
 :GHS05
 :GHS09
CAS Number:25895-60-7
Formula:CH₃BNNa
MDL Number:MFCD00003516