Organischer Referenzstandard, 1-Brom-4-fluorbenzol 25.000 µg/ml in Methanol, Supelco®
Catalog # 48800.
Supplier: Merck
CAS Number:
Specifications
- VE:1
- CAS Nr.:460-00-4
- Bezeichnung:1-Brom-4-fluorbenzol 25.000 µg/ml in Methanol
- Formel:C₆H₄BrF
- Bezeichnung:1-Bromo-4-fluorobenzene,