4-Bromchinolin
5 g
Katalognr. MOLE58511676-5G
Marketsource
Anbieter: Molekula
Direkt zu
Product Details & Documents
- CAS Nummer:3964-04-3
- Formel:C₉H₆BrN
- MDL-Nummer:MFCD07644514
- Molekulargewicht:208.06
- Lagertemperatur:Kühlschrank
- UN:3261
- ADR:8,II
Technische Daten
Technische Daten
- VE:5 g
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



