Acetophenon
Glass bottle for solids, 100 mg
PROOMM0237.01
: Methyl phenyl ketone, Phenyl methyl ketone
- :98-86-2
- :C₈H₈O
- :MFCD00008724
- :120.15 g/mol
- :13,00074
- :201,7 °C (1013 hPa)
- :20 °C
- :1,027 g/cm³ (20 °C)
- :82 °C
- :Raumtemperatur
- :202-708-7
- :3334
- :9,
- Packung:Glass bottle for solids
- VE:100 mg