Piston pump
Kolbenpumpe für wässrige und petrochemische Erzeugnisse, PP/PE, 450 ml/stroke, 840 mm
Katalognr. BURK5613-0030
Anbieter: BURKLE
Direkt zu
Product Details & Documents
Piston pump is used for watery and petrochemical products.
Pump capacity adjustable from 100, 250, 350 and 500 ml per stroke. The piston pump is attached to the container which is to be filled using a 2" or 3/4" barrel thread. After mounting, the pump can still be rotated around its axis so that containers to be filled can be placed on the barrel lid.
Technische Daten
- Förderleistung:450 ml/stroke
- Bezeichnung:Kolbenpumpe für wässrige und petrochemische Erzeugnisse
- Material:PP/PE
- Eintauch-Tiefe:840 mm