Schwefelsäure 50%
Supplier: VWR Collection
Certificates
About this item
CAS Number:7664-93-9
:
Formula:H₂SO₄
MDL Number:MFCD00064589
Boiling Pt:∼100 °C (1013 hPa)
Melting Pt:10.38
Density:1.1…1.4 g/cm³ (20 °C)
Molecular Weight:98.08 g/mol
Storage Temperature:Raumtemperatur
EINECS:231-639-5
UN:2796
ADR:8 II
:GHS05
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together