Natriumdodecylsulfat (SDS)
Catalog # MOLE10325163-1KG
Supplier: Molekula
CAS Number:
Specifications
- Packung:Glass bottle for solids
- VE:1 kg
- Seq:4024
Specifications
About this item
:
:288.38 g/mol
Merck Index:14,08636
Melting Pt:204…207 °C
Density:1.1 g/cm³ (20 °C)
Flash Pt:>100 °C
Storage Temperature:Raumtemperatur
EINECS:205-788-1
UN:1325
ADR:4.1,II
:GHS02
:GHS06
CAS Number:151-21-3
Formula:CH₃(CH₂)₁₁OSO₃Na
MDL Number:MFCD00036175