1-Chlorhexan
Catalog # EHERCA11416000
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Packung:Glasflasche
- VE:1 mL
- Seq:2070
Specifications
About this item
:
:120.62 g/mol
Merck Index:13,02163
Boiling Pt:∼135 °C (1013 hPa)
Melting Pt:–94 °C
Density:0.872 g/cm³ (20 °C)
Flash Pt:27 °C
Storage Temperature:Raumtemperatur
EINECS:208-859-5
UN:1993
ADR:3,III
CAS Number:544-10-5
Formula:C₆H₁₃Cl
MDL Number:MFCD00001018