Pelargonsäure
Glasflasche, 50 mL
Catalog # ROTH7015.1
Marketsource
Supplier: Roth Carl
Jump to:
Product Details & Documents
Synonyms: Nonanoic acid, Pelargonic Acid
- CAS Number:112-05-0
- Formula:C₉H₁₈O₂
- MDL Number:MFCD00004433
- Molecular Weight:158.24 g/mol
- Merck Index:12,07198
- Boiling Pt:253 °C (1013 hPa)
- Melting Pt:12 °C
- Density:0,9055 g/cm³ (20 °C)
- Flash Pt:133 °C
- Storage Temperature:Kühlschrank
- EINECS:203-931-2
- UN:3265
- ADR:8,III
Specifications
Specifications
- Packung:Glasflasche
- VE:50 mL