Arachinsäure
Glasflasche für Feststoffe, 5 g
Catalog # MOLE13447401-5G
Marketsource
Supplier: Molekula
Jump to:
Product Details & Documents
Synonyms: Eicosanylic acid, Arachidinic acid, Icosanoic acid, Eicosanoic acid
- CAS Number:506-30-9
- Formula:C₂₀H₄₀O₂
- MDL Number:MFCD00002755
- Molecular Weight:312.54 g/mol
- Merck Index:12,00802
- Boiling Pt:328 °C (1013 hPa)
- Melting Pt:75,5 °C
- Density:0,824 g/cm³ (100 °C) (Lit.)
- Storage Temperature:Gefrierschrank
- EINECS:208-031-3
Specifications
Specifications
- Packung:Glasflasche für Feststoffe
- VE:5 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



