Arachinsäure
Glasflasche für Feststoffe, 500 mg
Katalognr. MAYA1030
Anbieter: MATREYA
Direkt zu
Product Details & Documents
Synonyme: Eicosanylic acid, Arachidinic acid, Icosanoic acid, Eicosanoic acid
- CAS Nummer:506-30-9
- Formel:C₂₀H₄₀O₂
- MDL-Nummer:MFCD00002755
- Molekulargewicht:312.54 g/mol
- Merck-Index:12,00802
- Siedepunkt:328 °C (1013 hPa)
- Schmelzender Platingehalt:75,5 °C
- Dichte:0,824 g/cm³ (100 °C) (Lit.)
- Lagertemperatur:Gefrierschrank
- EINECS:208-031-3
Technische Daten
Technische Daten
- Packung:Glasflasche für Feststoffe
- VE:500 mg
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



