Amantadinsulfat, Sigma-Aldrich®
8.18383.0100
: Merck
:
- Packung:Glasflasche
- VE:100 g
- Seq:3401
- Solubility:(20 °C) insoluble
- Melting Point:299 - 301 °C
- Molar Mass:400.57 g/mol
- pH:4.9 (100 g/l, H2O, 20 °C) (slurry)
- Synonyms:1-Aminoadamantane sulfate, Adamantylamine sulfate
- Hill Formula:C20H36N2O4S
- Chemical Formula:C20H34N2 * H2SO4
:
:386.58 g/mol
:299…301 °C
:M-Clarity™ quality level = MQ200
:250-604-5
:31377-23-8
:C₁₀H₁₉NO₄S
:MFCD00077251