Order Entry
Switzerland
ContactUsLinkComponent
1,1,2,2-Tetrabromethan, Sigma-Aldrich®
 
1,1,2,2-Tetrabromethan, Sigma-Aldrich®
  1.00034.1000
 :  Merck
 :  
undefined
 
1,1,2,2-Tetrabromethan, Sigma-Aldrich®
  1.00034.1000
 :  Merck
 :  

 

  • Packung:
    Glasflasche
  • VE:
    1 L
  • Seq:
    5037

 

 

  • Solubility:
    0.68 g/l (25 °C)
  • Melting Point:
    -1 °C
  • Molar Mass:
    345.65 g/mol
  • Boiling Point:
    119 °C (20 hPa)
  • Vapor Pressure:
    0.03 - 0.13 hPa (20 °C)
  • Density:
    2.96 g/cm3 (25 °C)
  • Ignition Point:
    335 °C
  • Synonyms:
    TBE, Muthmanns liquid, Acetylene tetrabromide
  • Hill Formula:
    C2H2Br4
  • Chemical Formula:
    Br2CHCHBr2

 

 

 :
 :345.65 g/mol
 :13,09261
 :244 °C (1013 hPa)
 :–1 °C
 :2,9656 g/cm³ (20 °C)
 :Raumtemperatur
 :201-191-5
 :2504
 :6.1,III
 :M-Clarity™ Qualitätsstufe = MQ200
 :GHS06
 :79-27-6
 :Br₂HCCHBr₂
 :MFCD00000133